Please login to your online account to display your discounted pricing

Rat anti-Ly-6G, Clone: RB6-8C5, PE, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP200440500UG

 View all options for this product


For Research Use Only

Catalog No. NBP20044055


Description & Specifications


Antigen Ly-6G
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications In vivo Assay
Clone RB6-8C5
Concentration 0.2mg/mL
Conjugate PE
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen Gr1).
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 500μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 546644
Test Specificity Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.