Please login to your online account to display your discounted pricing

Rat anti-Ly-6G, Clone: RB6-8C5, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200440200UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20044026

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications In vivo Assay
Primary or Secondary Primary
Antigen Ly-6G
Format Affinity Purified
Host Species Rat
Immunogen Gr1).
Isotype IgG2b, κ
Quantity 200μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 546644
Test Specificity Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.
Concentration 0.2mg/mL
Purification Method Affinity Purified
Clone RB6-8C5
Conjugate PE
Cross Reactivity Mouse