Please login to your online account to display your discounted pricing

Rat anti-Ly-6G, Clone: RB6-8C5, PE-Cy5, Novus Biologicals™

$321.00 - $418.00

Specifications

Antigen Ly-6G
Clone RB6-8C5
Immunogen Gr1).
Gene ID (Entrez) 546644
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20016526

 
novus biologicals canada
NBP200165200UG
200μg Each for $418.00

NBP20016510

 
novus biologicals canada
NBP200165100UG
100μg Each for $321.00
Description & Specifications

Specifications

Antigen Ly-6G
Clone RB6-8C5
Immunogen Gr1).
Gene ID (Entrez) 546644
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry