Please login to your online account to display your discounted pricing

Rat anti-Ly-6G, Clone: RB6-8C5, PE-Cy5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200165100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20016510

  • / Each
 

Description & Specifications

Specifications

Antigen Ly-6G
Applications Flow Cytometry
Clone RB6-8C5
Conjugate PE-Cy5
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Immunogen Gr1).
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 546644
Test Specificity Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.