Please login to your online account to display your discounted pricing

Rat anti-Ly-6G, Clone: RB6-8C5, Novus Biologicals™

$252.00 - $321.00

Specifications

Antigen Ly-6G
Clone RB6-8C5
Concentration 0.5mg/mL
Immunogen Gr1).
Gene ID (Entrez) 546644
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20044110

 
novus biologicals canada
NBP200441100UG
100μg Each for $252.00

NBP20044155

 
novus biologicals canada
NBP200441500UG
500μg Each for $321.00
Description & Specifications

Specifications

Antigen Ly-6G
Clone RB6-8C5
Concentration 0.5mg/mL
Immunogen Gr1).
Gene ID (Entrez) 546644
Formulation Aqueous buffer containing carrier protein.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting