Please login to your online account to display your discounted pricing

Rat anti-Ly-6G, Clone: RB6-8C5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200441100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20044110

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Antigen Ly-6G
Format Affinity Purified
Formulation Aqueous buffer containing carrier protein.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 100μg
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Cross Reactivity Mouse
Host Species Rat
Immunogen Gr1).
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 546644
Test Specificity Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.
Clone RB6-8C5
Conjugate Unlabeled