Please login to your online account to display your discounted pricing

Rat anti-Ly-6A/E, Clone: D7, FITC, Novus Biologicals™

$171.00 - $354.00

Specifications

Antigen Ly-6A/E
Clone D7
Concentration 0.5mg/mL
Immunogen IL-2-dependent mouse T-cell line (CTL-L)
Gene ID (Entrez) 110454
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20038110

 
novus biologicals canada
NBP200381100UG
100μg Each for $171.00

NBP20038155

 
novus biologicals canada
NBP200381500UG
500μg Each for $354.00
Description & Specifications

Specifications

Antigen Ly-6A/E
Clone D7
Concentration 0.5mg/mL
Immunogen IL-2-dependent mouse T-cell line (CTL-L)
Gene ID (Entrez) 110454
Formulation Immunogen affinity purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry