Please login to your online account to display your discounted pricing

Rat anti-Ly-6A/E, Clone: D7, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200381500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20038155

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Quantity 500μg
Cross Reactivity Mouse
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Immunogen IL-2-dependent mouse T-cell line (CTL-L)
Antigen Ly-6A/E
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Format Affinity Purified
Concentration 0.5mg/mL
Gene ID (Entrez) 110454
Formulation Immunogen affinity purified
Host Species Rat
Conjugate FITC
Clone D7
Isotype IgG2a, κ
Purification Method Affinity Purified
Test Specificity Ly-6A/E (D7)