Please login to your online account to display your discounted pricing

Rat anti-Ly-6A/E, Clone: D7, Biotin, Novus Biologicals™

$128.00 - $321.00

Specifications

Antigen Ly-6A/E
Clone D7
Concentration 0.5mg/mL
Immunogen IL-2-dependent mouse T-cell line (CTL-L)
Gene ID (Entrez) 110454
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20044710

 
novus biologicals canada
NBP200447100UG
100μg Each for $128.00

NBP20044755

 
novus biologicals canada
NBP200447500UG
500μg Each for $321.00
Description & Specifications

Specifications

Antigen Ly-6A/E
Clone D7
Concentration 0.5mg/mL
Immunogen IL-2-dependent mouse T-cell line (CTL-L)
Gene ID (Entrez) 110454
Formulation Immunogen affinity purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry