Please login to your online account to display your discounted pricing

Rat anti-Ly-6A/E, Clone: D7, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200447500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20044755

  • / Each
 

Description & Specifications

Specifications

Antigen Ly-6A/E
Host Species Rat
Regulatory Status RUO
Conjugate Biotin
Cross Reactivity Mouse
Format Affinity Purified
Formulation Immunogen affinity purified
Applications Flow Cytometry
Immunogen IL-2-dependent mouse T-cell line (CTL-L)
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 500μg
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 110454
Test Specificity Ly-6A/E (D7)
Clone D7
Concentration 0.5mg/mL