Please login to your online account to display your discounted pricing

Rat anti-Ly-6A/E, Clone: D7, Biotin, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP200447100UG

 View all options for this product


For Research Use Only

Catalog No. NBP20044710


Description & Specifications


Antigen Ly-6A/E
Applications Flow Cytometry
Clone D7
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Format Affinity Purified
Formulation Immunogen affinity purified
Host Species Rat
Immunogen IL-2-dependent mouse T-cell line (CTL-L)
Isotype IgG2a, κ
Purification Method Affinity Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 110454
Test Specificity Ly-6A/E (D7)