Please login to your online account to display your discounted pricing

Rat anti-Ly-49A/D, Clone: 12A8, Novus Biologicals™

$285.00 - $362.00

Specifications

Antigen Ly-49A/D
Clone 12A8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is Ly-49A/D.
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20030010

 
novus biologicals canada
NBP200300100UG
100μg Each for $285.00

NBP20030055

 
novus biologicals canada
NBP200300500UG
500μg Each for $362.00
Description & Specifications

Specifications

Antigen Ly-49A/D
Clone 12A8
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is Ly-49A/D.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry