Please login to your online account to display your discounted pricing

Rat anti-Ly-49A/D, Clone: 12A8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200300500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20030055

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Host Species Rat
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Quantity 500μg
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Antigen Ly-49A/D
Cross Reactivity Mouse
Format Affinity Purified
Immunogen The immunogen for this antibody is Ly-49A/D.
Isotype IgG2a, κ
Test Specificity 12A8 does not react with BALB/c, DBA/2, AKR, or CBA/J Ly-D49D.
Clone 12A8
Regulatory Status RUO
Concentration 0.5mg/mL
Applications Flow Cytometry