Please login to your online account to display your discounted pricing

Rat anti-Ly-49A/D, Clone: 12A8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200300100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20030010

  • / Each
 

Description & Specifications

Specifications

Antigen Ly-49A/D
Format Affinity Purified
Host Species Rat
Primary or Secondary Primary
Applications Flow Cytometry
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Quantity 100μg
Purification Method Affinity Purified
Immunogen The immunogen for this antibody is Ly-49A/D.
Conjugate Unlabeled
Storage Requirements Store at 4°C. Do not freeze.
Regulatory Status RUO
Isotype IgG2a, κ
Concentration 0.5mg/mL
Clone 12A8
Test Specificity 12A8 does not react with BALB/c, DBA/2, AKR, or CBA/J Ly-D49D.