Please login to your online account to display your discounted pricing

Rat anti-Lgl1, Clone: 4D11, Novus Biologicals™

$285.00 - $362.00

Specifications

Antigen Lgl1
Clone 4D11
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is LGL1.
Gene ID (Entrez) 3996
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20029510

 
novus biologicals canada
NBP200295100UG
100μg Each for $285.00

NBP20029555

 
novus biologicals canada
NBP200295500UG
500μg Each for $362.00
Description & Specifications

Specifications

Antigen Lgl1
Clone 4D11
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is LGL1.
Gene ID (Entrez) 3996
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunoprecipitation