Please login to your online account to display your discounted pricing

Rat anti-Lgl1, Clone: 4D11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200295100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20029510

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Immunogen The immunogen for this antibody is LGL1.
Isotype IgG2a, κ
Conjugate Unlabeled
Applications Flow Cytometry
Applications Immunoprecipitation
Antigen Lgl1
Purification Method Affinity Purified
Quantity 100μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3996
Clone 4D11
Concentration 0.5mg/mL
Format Affinity Purified
Cross Reactivity Mouse