Please login to your online account to display your discounted pricing

Rat anti-Lgl1, Clone: 4D11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200295500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20029555

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunoprecipitation
Immunogen The immunogen for this antibody is LGL1.
Quantity 500μg
Cross Reactivity Mouse
Format Affinity Purified
Host Species Rat
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3996
Conjugate Unlabeled
Antigen Lgl1
Isotype IgG2a, κ
Clone 4D11
Concentration 0.5mg/mL
Purification Method Affinity Purified
Primary or Secondary Primary