Please login to your online account to display your discounted pricing

Rat anti-LAMP-2/CD107b, Clone: GL2A7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19749902ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19749902

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen LAMP-2/CD107b
Format Ascites
Formulation Liquid. Rat Ascites.
Gene Accession No. P13473
Host Species Rat
Immunogen Mouse liver lysosomal membrane preparation.
Isotype IgG
Quantity 0.2mL
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3920
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Purification Method Ascites
Clone GL2A7
Conjugate Unlabeled