Please login to your online account to display your discounted pricing

Rat anti-L-Selectin/CD62L, Clone: MEL-14, PE-Cy5, Novus Biologicals™

$296.00 - $428.00

Specifications

Antigen L-Selectin/CD62L
Clone MEL-14
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is L-selectin.
Gene ID (Entrez) 6402
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20026110

 
novus biologicals canada
NBP200261100UG
100μg Each for $296.00

NBP20026126

 
novus biologicals canada
NBP200261200UG
200μg Each for $428.00
Description & Specifications

Specifications

Antigen L-Selectin/CD62L
Clone MEL-14
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is L-selectin.
Gene ID (Entrez) 6402
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate PE-Cy5
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry