Please login to your online account to display your discounted pricing

Rat anti-L-Selectin/CD62L, Clone: MEL-14, Novus Biologicals™

$362.00 - $362.00

Specifications

Antigen L-Selectin/CD62L
Clone MEL-14
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is L-selectin.
Gene ID (Entrez) 6402
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20026055

 
novus biologicals canada
NBP200260500UG
500μg Each for $362.00
Description & Specifications

Specifications

Antigen L-Selectin/CD62L
Clone MEL-14
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is L-selectin.
Gene ID (Entrez) 6402
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation