Please login to your online account to display your discounted pricing

Rat anti-L-Selectin/CD62L, Clone: MEL-14, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200260500UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20026055

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Conjugate Unlabeled
Antigen L-Selectin/CD62L
Cross Reactivity Mouse
Primary or Secondary Primary
Purification Method Affinity Purified
Host Species Rat
Clone MEL-14
Isotype IgG2a, κ
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is L-selectin.
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6402
Format Affinity Purified
Quantity 500μg