Please login to your online account to display your discounted pricing

Rat anti-L-Selectin/CD62L, Clone: MEL-14, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200260100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20026010

  • / Each
 

Description & Specifications

Specifications

Host Species Rat
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Storage Requirements Store at 4°C. Do not freeze.
Antigen L-Selectin/CD62L
Format Affinity Purified
Immunogen The immunogen for this antibody is L-selectin.
Isotype IgG2a, κ
Quantity 100μg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6402
Clone MEL-14
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Purification Method Affinity Purified
Primary or Secondary Primary