Please login to your online account to display your discounted pricing

Rat anti-L-Selectin/CD62L, Clone: MEL-14, Biotin, Novus Biologicals™

$131.00 - $395.00

Specifications

Antigen L-Selectin/CD62L
Clone MEL-14
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is L-selectin.
Gene ID (Entrez) 6402
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14362505

 
novus biologicals canada
NBP143625005MG
0.05mg Each for $131.00

NBP14362501

 
novus biologicals canada
NBP14362501MG
0.1mg Each for $197.00

NBP1436255

 
novus biologicals canada
NBP14362505MG
0.5mg Each for $395.00
Description & Specifications

Specifications

Antigen L-Selectin/CD62L
Clone MEL-14
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody is L-selectin.
Gene ID (Entrez) 6402
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry