Please login to your online account to display your discounted pricing

Rat anti-Integrin beta 7, Clone: FIB504, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen Integrin beta 7
Clone FIB504
Concentration 0.5mg/mL
Immunogen TK1 cells
Gene ID (Entrez) 3695
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14353801

 
novus biologicals canada
NBP14353801MG
0.1mg Each for $329.00

NBP14353803

 
novus biologicals canada
NBP1435380025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen Integrin beta 7
Clone FIB504
Concentration 0.5mg/mL
Immunogen TK1 cells
Gene ID (Entrez) 3695
Formulation Aqueous buffer (contains stabilizer if necessary). Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Baboon
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry