Please login to your online account to display your discounted pricing

Rat anti-Integrin beta 4/CD104, Clone: 439-9B, Biotin, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen Integrin beta 4/CD104
Clone 439-9B
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD104.
Gene Accession No. P16144
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14359901

 
novus biologicals canada
NBP14359901MG
0.1mg Each for $252.00

NBP14359903

 
novus biologicals canada
NBP1435990025MG
0.025mg Each for $131.00
Description & Specifications

Specifications

Antigen Integrin beta 4/CD104
Clone 439-9B
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD104.
Gene Accession No. P16144
Gene ID (Entrez) 3691
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2b
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)