missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-Integrin beta 1/CD29, Clone: HMb1-1, Novus Biologicals™

$285.00 - $472.00

Specifications

Antigen Integrin beta 1/CD29
Clone HMb1-1
Concentration 1mg/mL
Immunogen The immunogen for this antibody was CD29.
Gene ID (Entrez) 3688
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP2332025 Novus Biologicals Canada
NBP23320205MG
0.5mg Each for $472.00
NBP2332025M Novus Biologicals Canada
NBP233202005MG
0.05mg Each for $285.00
Specifications Description & Specifications

Specifications

Integrin beta 1/CD29
1mg/mL
3688
Monoclonal
Store at 4°C. Do not freeze.
IgG
Affinity Purified
Flow Cytometry,Functional Assay
HMb1-1
The immunogen for this antibody was CD29.
Aqueous Buffer. Immunogen affinity purified.
Primary
Rat
Unlabeled
Affinity Purified