Please login to your online account to display your discounted pricing

Rat anti-Integrin beta 1/CD29, Clone: HMb1-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233202005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2332025M

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Mouse
Immunogen The immunogen for this antibody was CD29.
Regulatory Status RUO
Formulation Aqueous Buffer. Immunogen affinity purified.
Gene ID (Entrez) 3688
Concentration 1mg/mL
Format Affinity Purified
Isotype IgG
Quantity 0.05mg
Antigen Integrin beta 1/CD29
Purification Method Affinity Purified
Clone HMb1-1
Primary or Secondary Primary