Please login to your online account to display your discounted pricing

Rat anti-Integrin beta 1/CD29, Clone: HMb1-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23320205MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2332025

  • / Each
 

Description & Specifications

Specifications

Antigen Integrin beta 1/CD29
Applications Flow Cytometry
Applications Functional Assay
Clone HMb1-1
Concentration 1mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation Aqueous Buffer. Immunogen affinity purified.
Host Species Rat
Immunogen The immunogen for this antibody was CD29.
Isotype IgG
Purification Method Affinity Purified
Quantity 0.5mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3688
Target Species Murine