Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha M/CD11b, Clone: M1/70, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Integrin alpha M/CD11b
Clone M1/70
Concentration 0.2mg/mL
Immunogen B10 mouse spleen cells enriched for T lymphocytes
Gene Accession No. P11215
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23426301

 
novus biologicals canada
NBP23426301ML
0.1mL Each for $139.00

NBP23426305

 
novus biologicals canada
NBP23426305ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Integrin alpha M/CD11b
Clone M1/70
Concentration 0.2mg/mL
Immunogen B10 mouse spleen cells enriched for T lymphocytes
Gene Accession No. P11215
Gene ID (Entrez) 3684
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Rat
Cross Reactivity Baboon
Cross Reactivity Chimpanzee
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rhesus Ma
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation