Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha M/CD11b, Clone: M1/70, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426305

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P11215
Host Species Rat
Antigen Integrin alpha M/CD11b
Concentration 0.2mg/mL
Primary or Secondary Primary
Cross Reactivity Baboon
Cross Reactivity Chimpanzee
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rhesus Ma
Immunogen B10 mouse spleen cells enriched for T lymphocytes
Isotype IgG2b, κ
Conjugate Unlabeled
Gene ID (Entrez) 3684
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Quantity 0.5mL
Clone M1/70
Reconstitution Protein A purified
Purification Method Purified
Format Purified
Regulatory Status RUO