Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha M/CD11b, Clone: M1/70, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23426301ML

 View all options for this product


For Research Use Only

Catalog No. NBP23426301


Description & Specifications


Antigen Integrin alpha M/CD11b
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone M1/70
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Baboon
Cross Reactivity Chimpanzee
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rhesus Ma
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P11215
Host Species Rat
Immunogen B10 mouse spleen cells enriched for T lymphocytes
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3684
Reconstitution Protein A purified