Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha M/CD11b, Clone: M1/70, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467802

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG2b, κ
Quantity 0.2mg
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Baboon
Cross Reactivity Chimpanzee
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rhesus Ma
Antigen Integrin alpha M/CD11b
Host Species Rat
Immunogen B10 mouse spleen cells enriched for T lymphocytes
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Protein A purified
Clone M1/70
Concentration 1mg/mL
Gene Accession No. P11215
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3684