Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha M/CD11b, Clone: M1/70, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23467801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23467801

  • / Each
 

Description & Specifications

Specifications

Antigen Integrin alpha M/CD11b
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen B10 mouse spleen cells enriched for T lymphocytes
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Cross Reactivity Baboon
Cross Reactivity Chimpanzee
Cross Reactivity Cynomolgus
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rabbit
Cross Reactivity Rhesus Ma
Format Purified
Gene Accession No. P11215
Host Species Rat
Isotype IgG2b, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3684
Clone M1/70
Concentration 1mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Reconstitution Protein A purified