Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha L/CD11a, Clone: M17/4, PE, Novus Biologicals™

$246.00 - $354.00

Specifications

Antigen Integrin alpha L/CD11a
Clone M17/4
Concentration 0.2mg/mL
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Gene ID (Entrez) 3683
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20004410

 
novus biologicals canada
NBP200044100UG
100μg Each for $246.00

NBP20004426

 
novus biologicals canada
NBP200044200UG
200μg Each for $354.00
Description & Specifications

Specifications

Antigen Integrin alpha L/CD11a
Clone M17/4
Concentration 0.2mg/mL
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Gene ID (Entrez) 3683
Formulation IgG purified
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate PE
Format Purified
Purification Method Purified
Applications Flow Cytometry