Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha L/CD11a, Clone: M17/4, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200044100UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20004410

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Regulatory Status RUO
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Format Purified
Storage Requirements Store at 4°C. Do not freeze.
Conjugate PE
Monoclonal or Polyclonal Monoclonal
Formulation IgG purified
Concentration 0.2mg/mL
Primary or Secondary Primary
Quantity 100μg
Host Species Rat
Test Specificity The rat monoclonal antibody M17/4 reacts with CD11a (alpha-subunit of murine LFA-1), a 180kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.
Isotype IgG2a
Clone M17/4
Purification Method Purified
Antigen Integrin alpha L/CD11a
Gene ID (Entrez) 3683
Applications Flow Cytometry