Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha L/CD11a, Clone: M17/4, PE, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP200044200UG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP20004426

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Mouse
Antigen Integrin alpha L/CD11a
Format Purified
Host Species Rat
Applications Flow Cytometry
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Quantity 200μg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 3683
Conjugate PE
Formulation IgG purified
Isotype IgG2a
Purification Method Purified
Monoclonal or Polyclonal Monoclonal
Test Specificity The rat monoclonal antibody M17/4 reacts with CD11a (alpha-subunit of murine LFA-1), a 180kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.
Clone M17/4
Concentration 0.2mg/mL