Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha L/CD11a, Clone: M17/4, Biotin, Novus Biologicals™

$96.00 - $321.00

Specifications

Antigen Integrin alpha L/CD11a
Clone M17/4
Concentration 0.5mg/mL
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Gene ID (Entrez) 3683
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14367901

 
novus biologicals canada
NBP14367901MG
0.1mg Each for $171.00

NBP14367905

 
novus biologicals canada
NBP143679005MG
0.05mg Each for $96.00

NBP1436795

 
novus biologicals canada
NBP14367905MG
0.5mg Each for $321.00
Description & Specifications

Specifications

Antigen Integrin alpha L/CD11a
Clone M17/4
Concentration 0.5mg/mL
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Gene ID (Entrez) 3683
Formulation Phosphate buffer pH 7.2. IgG purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a
Conjugate Biotin
Format Purified
Purification Method Purified
Applications Flow Cytometry