Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha L/CD11a, Clone: M17/4, Biotin, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143679005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14367905

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Flow Cytometry
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Isotype IgG2a
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3683
Conjugate Biotin
Cross Reactivity Mouse
Antigen Integrin alpha L/CD11a
Formulation Phosphate buffer pH 7.2. IgG purified.
Host Species Rat
Purification Method Purified
Storage Requirements Store at 4°C. Do not freeze.
Monoclonal or Polyclonal Monoclonal
Test Specificity The rat monoclonal antibody M17/4 reacts with CD11a (alpha-subunit of murine LFA-1), a 180kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.
Clone M17/4
Concentration 0.5mg/mL