Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha L/CD11a, Clone: M17/4, Biotin, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP14367901MG

 View all options for this product


For Research Use Only

Catalog No. NBP14367901


Description & Specifications


Antigen Integrin alpha L/CD11a
Applications Flow Cytometry
Clone M17/4
Concentration 0.5mg/mL
Conjugate Biotin
Cross Reactivity Mouse
Format Purified
Formulation Phosphate buffer pH 7.2. IgG purified.
Host Species Rat
Immunogen C57BL/6 mouse splenic secondary cytotoxic T lymphocytes
Isotype IgG2a
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3683
Test Specificity The rat monoclonal antibody M17/4 reacts with CD11a (alpha-subunit of murine LFA-1), a 180kDa type I transmembrane glycoprotein expressed on B and T lymphocytes, monocytes, macrophages, neutrophils, basophils and eosinophils.