Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 6/CD49f, Clone: GoH3, Novus Biologicals™

$252.00 - $340.00

Specifications

Antigen Integrin alpha 6/CD49f
Clone GoH3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD49f.
Gene Accession No. P23229
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14340001

 
novus biologicals canada
NBP14340001MG
0.1mg Each for $285.00

NBP14340005

 
novus biologicals canada
NBP143400005MG
0.05mg Each for $252.00

NBP1434005

 
novus biologicals canada
NBP14340005MG
0.5mg Each for $340.00
Description & Specifications

Specifications

Antigen Integrin alpha 6/CD49f
Clone GoH3
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD49f.
Gene Accession No. P23229
Gene ID (Entrez) 3655
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation