Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 6/CD49f, Clone: GoH3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14340005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1434005

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen The immunogen for this antibody was CD49f.
Formulation PBS [pH 7.2]
Host Species Rat
Isotype IgG2a, κ
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Integrin alpha 6/CD49f
Format Affinity Purified
Gene Accession No. P23229
Quantity 0.5mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3655
Test Specificity May also react with Bovine.
Clone GoH3
Concentration 0.5mg/mL