Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 6/CD49f, Clone: GoH3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP143400005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14340005

  • / Each
 

Description & Specifications

Specifications

Antigen Integrin alpha 6/CD49f
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS [pH 7.2]
Gene Accession No. P23229
Host Species Rat
Immunogen The immunogen for this antibody was CD49f.
Purification Method Affinity Purified
Quantity 0.05mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3655
Clone GoH3
Concentration 0.5mg/mL
Conjugate Unlabeled
Isotype IgG2a, κ
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity May also react with Bovine.