Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 4/CD49d, Clone: R1-2, PE, Novus Biologicals™

$246.00 - $386.00

Specifications

Antigen Integrin alpha 4/CD49d
Clone R1-2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is Integrin alpha 4.
Gene ID (Entrez) 3676
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP20023610

 
novus biologicals canada
NBP200236100UG
100μg Each for $246.00

NBP20023626

 
novus biologicals canada
NBP200236200UG
200μg Each for $386.00
Description & Specifications

Specifications

Antigen Integrin alpha 4/CD49d
Clone R1-2
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody is Integrin alpha 4.
Gene ID (Entrez) 3676
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2b, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry