Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 4 beta 7/LPAM-1, Clone: DATK32, Novus Biologicals™

$219.00 - $362.00

Specifications

Antigen Integrin alpha 4 beta 7/LPAM-1
Clone DATK32
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was LPAM1.
Gene ID (Entrez) 3676
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14328301

 
novus biologicals canada
NBP14328301MG
0.1mg Each for $274.00

NBP14328305

 
novus biologicals canada
NBP143283005MG
0.05mg Each for $219.00

NBP1432835

 
novus biologicals canada
NBP14328305MG
0.5mg Each for $362.00
Description & Specifications

Specifications

Antigen Integrin alpha 4 beta 7/LPAM-1
Clone DATK32
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was LPAM1.
Gene ID (Entrez) 3676
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay