Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 4 beta 7/LPAM-1, Clone: DATK32, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328305MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1432835

  • / Each
 

Description & Specifications

Specifications

Antigen Integrin alpha 4 beta 7/LPAM-1
Applications Flow Cytometry
Applications Functional Assay
Gene ID (Entrez) 3676
Cross Reactivity Mouse
Format Affinity Purified
Formulation Phosphate buffer pH 7.2
Host Species Rat
Immunogen The immunogen for this antibody was LPAM1.
Quantity 0.5mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Conjugate Unlabeled
Isotype IgG2a, κ
Purification Method Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Clone DATK32