Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 4 beta 7/LPAM-1, Clone: DATK32, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14328301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14328301

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Cross Reactivity Mouse
Quantity 0.1mg
Immunogen The immunogen for this antibody was LPAM1.
Conjugate Unlabeled
Antigen Integrin alpha 4 beta 7/LPAM-1
Purification Method Affinity Purified
Monoclonal or Polyclonal Monoclonal
Host Species Rat
Gene ID (Entrez) 3676
Regulatory Status RUO
Formulation Phosphate buffer pH 7.2
Format Affinity Purified
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Clone DATK32
Concentration 0.5mg/mL
Isotype IgG2a, κ