missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Rat anti-Integrin alpha 2b/CD41, Clone: MWReg30, Novus Biologicals™

$274.00 - $406.00

Specifications

Antigen Integrin alpha 2b/CD41
Clone MWReg30
Immunogen The immunogen for this antibody was CD41.
Gene ID (Entrez) 3674
Formulation PBS [pH 7.2]
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP14341501 Novus Biologicals Canada
NBP14341501MG
0.1mg Each for $329.00
NBP14341505 Novus Biologicals Canada
NBP143415005MG
0.05mg Each for $274.00
NBP1434155 Novus Biologicals Canada
NBP14341505MG
0.5mg Each for $406.00
Specifications Description & Specifications

Specifications

Integrin alpha 2b/CD41
The immunogen for this antibody was CD41.
PBS [pH 7.2]
Primary
Rat
Unlabeled
Affinity Purified
MWReg30
3674
Monoclonal
Store at 4°C. Do not freeze.
IgG1, κ
Affinity Purified
ELISA,Flow Cytometry,Functional Assay,Immunoprecipitation