Please login to your online account to display your discounted pricing

Rat anti-IL-21 R, Clone: 4A9, Novus Biologicals™

$329.00 - $428.00

Specifications

Antigen IL-21 R
Clone 4A9
Immunogen The immunogen for this antibody was IL21R.
Gene ID (Entrez) 50615
Formulation Phosphate buffer pH 7.2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14335601

 
novus biologicals canada
NBP14335601MG
0.1mg Each for $329.00

NBP1433565

 
novus biologicals canada
NBP14335605MG
0.5mg Each for $428.00
Description & Specifications

Specifications

Antigen IL-21 R
Clone 4A9
Immunogen The immunogen for this antibody was IL21R.
Gene ID (Entrez) 50615
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry