Please login to your online account to display your discounted pricing

Rat anti-IL-21 R, Clone: 4A9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14335605MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP1433565

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Cross Reactivity Mouse
Monoclonal or Polyclonal Monoclonal
Antigen IL-21 R
Format Affinity Purified
Regulatory Status RUO
Formulation Phosphate buffer pH 7.2
Primary or Secondary Primary
Clone 4A9
Conjugate Unlabeled
Immunogen The immunogen for this antibody was IL21R.
Gene ID (Entrez) 50615
Purification Method Affinity Purified
Host Species Rat
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.5mg
Isotype IgG2a, κ