Please login to your online account to display your discounted pricing

Rat anti-IL-21 R, Clone: 4A9, Biotin, Novus Biologicals™

$175.00 - $549.00

Specifications

Antigen IL-21 R
Clone 4A9
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL21R.
Gene ID (Entrez) 50615
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14358705

 
novus biologicals canada
NBP143587005MG
0.05mg Each for $175.00

NBP14358701

 
novus biologicals canada
NBP14358701MG
0.1mg Each for $252.00

NBP1435875

 
novus biologicals canada
NBP14358705MG
0.5mg Each for $549.00
Description & Specifications

Specifications

Antigen IL-21 R
Clone 4A9
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was IL21R.
Gene ID (Entrez) 50615
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Rat
Cross Reactivity Mouse
Isotype IgG2a, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry